missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1H-Pyrrole-2-carbaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
Specifications
CAS | 1003-29-8 |
Assay Percent Range | 97% |
Molecular Formula | C5H5NO |
Quantity | 1 g |
Synonym | pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde |
InChI Key | ZSKGQVFRTSEPJT-UHFFFAOYSA-N |
SMILES | C1=CNC(=C1)C=O |
IUPAC Name | 1H-pyrrole-2-carbaldehyde |
Molecular Weight (g/mol) | 95.101 |
PubChem CID | 13854 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'