Learn More
Ammonium Acetate BAKER ANALYZED Reagent 1kg
Brand: JT Baker 0011.1000
142.50 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Ammonium Acetate BAKER ANALYZED Reagent 1kg
Code : NEW
Additional Details : CAS Number : 631-61-8 Weight : 1.00000kg