Learn More
1LT Buffer Standard pH 10.00 25oC ISO 17034 CRM Certified Reference Material
Brand: Reagecon CRM110025
1LT Buffer Standard pH 10.00 25oC ISO 17034 CRM Certified Reference Material
Additional Details : CAS Number : 1330-43-4 Weight : 1.06000kg
Safety and Handling
- BUFFER STANDARD pH 10.00 at 25°C ISO 17034 CERTIFIED REFERENCE MATERIAL
Signal Word
- Danger
Hazard Category
- Reproductive toxicity Category 1B
Hazard Statement
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : boric acid, disodiumsalt