Learn More
Thermo Scientific™ 1M TEAB and 50% Hydroxylamine Buffers for TMT Experiments
Common TMT buffers such as 1M Triethylammonium bicarbonate (TEAB) and 50% Hydroxylamine for isobaric mass tag labeling experiments.
Brand: Thermo Scientific™ 90114
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Pre-formulated TEAB and 50% hydroxylamine designed for use in TMT labeling and quenching of peptides.
Triethylamonium bicarbonate (TEAB) is a 10X dissolution buffer for isobaric mass tag labeling experiments. 50% hydroxylamine is a 10X quenching reagent for amine-reactive labeling for TMT experiments.
Specifications
1M Triethylammonium bicarbonate (TEAB) for TMT experiments | |
TMT™ | |
Cell Lysate | |
50 mL |
Buffer | |
Store protected from direct sunlight in a dry, cool, and well-ventilated area. | |
Protein Labeling |
Safety and Handling
- 1M TEAB and 50% Hydroxylamine Buffers for TMT Experiments
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H290-May be corrosive to metals.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H373-May cause damage to organs.
- H400-Very toxic to aquatic life.
Precautionary Statement
- P102-Keep out of reach of children.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hydroxylamine