missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1R,4S)-4-Aminocyclopent-2-enecarboxylic acid, 95%, 98% ee, Thermo Scientific™
Click to view available options
Quantity:
1 g
Packaging:
Glass Bottle
Specifications
Specifications
Melting Point | 180°C |
CAS | 134003-04-6 |
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Color | Brown |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Molecular Formula | C6H9NO2 |
Packaging | Glass Bottle |
MDL Number | MFCD00211285 |
Show More |
Safety and Handling
Product Identifier
- (1R,4S)-4-Aminocyclopent-2-enecarboxylic acid
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction