Learn More
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine, 97%, Thermo Scientific Chemicals
£19.90 - £430.00
Chemical Identifiers
CAS | 29841-69-8 |
---|---|
Molecular Formula | C14H16N2 |
Molecular Weight (g/mol) | 212.296 |
MDL Number | MFCD00082751 |
InChI Key | PONXTPCRRASWKW-KBPBESRZSA-N |
Synonym | 1s,2s---1,2-diphenylethylenediamine, 1s,2s---1,2-diphenyl-1,2-ethanediamine, 1s,2s-1,2-diphenylethane-1,2-diamine, 1s,2s-1,2-diphenyl-1,2-ethanediamine, 1s,2s---1,2-diamino-1,2-diphenylethane, s,s-dpen, 1,2-ethanediamine, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethane diamine, --1,2-diphenylethylenediamine, diphenylethylenediamine |
PubChem CID | 6931238 |
IUPAC Name | (1S,2S)-1,2-diphenylethane-1,2-diamine |
SMILES | C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337826
|
Thermo Scientific Alfa Aesar
L12968.MD |
250 mg |
£19.90
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327826
|
Thermo Scientific Alfa Aesar
L12968.03 |
1 g |
£38.30
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15478969
|
Thermo Scientific Alfa Aesar
L12968.06 |
5 g |
£126.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15488969
|
Thermo Scientific Alfa Aesar
L12968.14 |
25 g |
£430.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine solvation agent, For synthesis of enantiopure ethylenediamines by chirality transfer (condensation with diketones followed by reductive cleavage), Co-catalyst in the Ru catalyzed enantioselective hydrogenation of aromatic ketones, Versatile ligand for the formation of metal complexes.1 Used in the synthesis of chiral tropocoronands which have potential utility in asymmetric catalysis
Solubility
Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to air.
Chemical Identifiers
29841-69-8 | |
212.296 | |
PONXTPCRRASWKW-KBPBESRZSA-N | |
6931238 | |
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N |
C14H16N2 | |
MFCD00082751 | |
1s,2s---1,2-diphenylethylenediamine, 1s,2s---1,2-diphenyl-1,2-ethanediamine, 1s,2s-1,2-diphenylethane-1,2-diamine, 1s,2s-1,2-diphenyl-1,2-ethanediamine, 1s,2s---1,2-diamino-1,2-diphenylethane, s,s-dpen, 1,2-ethanediamine, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethane diamine, --1,2-diphenylethylenediamine, diphenylethylenediamine | |
(1S,2S)-1,2-diphenylethane-1,2-diamine |
Specifications
29841-69-8 | |
C14H16N2 | |
UN3259 | |
Air sensitive | |
Insoluble in water. | |
−102° (c=1 in Methanol) | |
(1S,2S)-1,2-diphenylethane-1,2-diamine | |
6931238 | |
97% |
80°C to 84°C | |
MFCD00082751 | |
3201645 | |
1s,2s---1,2-diphenylethylenediamine, 1s,2s---1,2-diphenyl-1,2-ethanediamine, 1s,2s-1,2-diphenylethane-1,2-diamine, 1s,2s-1,2-diphenyl-1,2-ethanediamine, 1s,2s---1,2-diamino-1,2-diphenylethane, s,s-dpen, 1,2-ethanediamine, 1,2-diphenyl-, 1s,2s, 1s,2s---1,2-diphenyl-1,2-ethane diamine, --1,2-diphenylethylenediamine, diphenylethylenediamine | |
PONXTPCRRASWKW-KBPBESRZSA-N | |
C1=CC=C(C=C1)C(C(C2=CC=CC=C2)N)N | |
212.296 | |
212.3 | |
(1S,2S)-(-)-1,2-Diphenyl-1,2-ethanediamine |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, SOLID, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only