missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 11358996
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132747-20-7
Molecular Formula C5H12Br2N2
Molecular Weight (g/mol) 259.97
MDL Number MFCD08272741
InChI Key ISYQWKOXKGJREA-UHFFFAOYNA-N
Synonym 1s,4s-2,5-diazabicyclo 2.2.1 heptane dihydrobromide, 1s,4s-+-2,5-diazabicyclo 2.2.1 heptane dihydrobromide, 1s,4s-2,5-diazabicylo 2.2.1 heptane dihydrobromide, 2,5-diazabicyclo 2.2.1 heptane,hydrobromide 1:2, 1s,4s-2,5-diazabicyclo 2.2.1 heptane dihydrobrom, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane dihydrobromide, 1s,2s-2,5-diazabicyclo 2.2.1 heptane dihydrobromide
PubChem CID 14612497
SMILES Br.Br.C1NC2CNC1C2

Specifications

Melting Point ∽290°C (decomposition)
Quantity 1 g
Beilstein 4258260
Sensitivity Hygroscopic
Optical Rotation +22° (c=1 in Water)
IUPAC Name (1S,4S)-2,5-diazabicyclo[2.2.1]heptane;dihydrobromide
Formula Weight 259.97
Percent Purity 98%
Chemical Name or Material (1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'