missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide, 98%

Product Code. p-7048058
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11368996

Brand: Thermo Scientific Alfa Aesar L20170.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 132747-20-7 | C5H12Br2N2 | 259.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132747-20-7
Molecular Formula C5H12Br2N2
Molecular Weight (g/mol) 259.97
MDL Number MFCD08272741
InChI Key ISYQWKOXKGJREA-UHFFFAOYNA-N
Synonym 1s,4s-2,5-diazabicyclo 2.2.1 heptane dihydrobromide, 1s,4s-+-2,5-diazabicyclo 2.2.1 heptane dihydrobromide, 1s,4s-2,5-diazabicylo 2.2.1 heptane dihydrobromide, 2,5-diazabicyclo 2.2.1 heptane,hydrobromide 1:2, 1s,4s-2,5-diazabicyclo 2.2.1 heptane dihydrobrom, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane dihydrobromide, 1s,2s-2,5-diazabicyclo 2.2.1 heptane dihydrobromide
PubChem CID 14612497
SMILES Br.Br.C1NC2CNC1C2

Specifications

Melting Point ∽290°C (decomposition)
Quantity 250 mg
Beilstein 4258260
Sensitivity Hygroscopic
Optical Rotation +22° (c=1 in Water)
IUPAC Name 2,5-diazabicyclo[2.2.1]heptane dihydrobromide
Formula Weight 259.97
Percent Purity 98%
Chemical Name or Material (1S,2S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.