missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,2S)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, 96%, Thermo Scientific Chemicals
£36.10 - £349.00
Chemical Identifiers
CAS | 2964-48-9 |
---|---|
Molecular Formula | C9H12N2O4 |
Molecular Weight (g/mol) | 212.2 |
MDL Number | MFCD00007359 |
InChI Key | OCYJXSUPZMNXEN-IUCAKERBSA-N |
PubChem CID | 224171 |
SMILES | C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10598761
|
Thermo Scientific Acros
334800050 |
5 g | Glass bottle |
£36.10
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10120611
|
Thermo Scientific Acros
334800250 |
25 g | Glass bottle |
£121.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10320944
|
Thermo Scientific Acros
334801000 |
100 g | Glass bottle |
£349.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2964-48-9 | |
212.2 | |
OCYJXSUPZMNXEN-IUCAKERBSA-N | |
C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-] |
C9H12N2O4 | |
MFCD00007359 | |
224171 |
Specifications
2964-48-9 | |
100.0 | |
Yellow to Beige | |
96% | |
MFCD00007359 | |
1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propanediol, 1s,2s-2-amino-1-4-nitrophenyl propane-1,3-diol, 1s,2s-2-amino-1-4-nitrophenyl-1,3-propanediol, 2-amino-1-4-nitro-phenyl-propane-1,3-diol, 1,3-propanediol, 2-amino-1-4-nitrophenyl-, 1s,2s, threomine, 1s,2s-2-azanyl-1-4-nitrophenyl propane-1,3-diol, 1s,2s-+-2-amino-1-4-nitrophenyl-1,3-propaned, 1,3-propanediol,2-amino-1-4-nitrophenyl-, 1s,2s, l-threo-+-2-amino-1-4-nitrophenyl-1,3-propanediol | |
C1=CC(=CC=C1C(C(CO)N)O)[N+](=O)[O-] | |
224171 | |
96% | |
(1S, 2S)-2-Amino-1-(4-nitrophenyl)propane-1, 3-diol |
95.0 | |
160°C to 166°C | |
Authentic | |
C9H12N2O4 | |
+ 31 | |
OCYJXSUPZMNXEN-IUCAKERBSA-N | |
212.2 | |
212.2 | |
Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 221-001-4
RUO – Research Use Only