missing translation for 'onlineSavingsMsg'
Learn More

(1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine, 97%, Thermo Scientific™

Product Code. 15498306
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a intermediate for pharmaceutical and asymmetirc synthesis.

Solubility
Insoluble in water.

Notes
Air Sensitive, store away from air. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 174677-83-9
Molecular Formula C44H44N2P2
Molecular Weight (g/mol) 662.80
MDL Number MFCD17014022
InChI Key OBHPYVNBXWUKNY-SQWPCXIINA-N
Synonym 1s,2s-n1,n1-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine, 1s,2s-n,n-bis 2-diphenylphosphino benzyl cyclohexane-1,2-diamine, 1s,2s-n1,n1-bis 2-diphenylphosphanyl phenyl methyl cyclohexane-1,2-diamine, 1s,2s-n,n'-bis 2-diphenylphosphino phenyl methyl-1,2-cyclohexanediamine, 1s,2s-n∼1∼,n∼1∼-bis 2-diphenylphosphanyl phenyl methyl cyclohexane-1,2-diamine
PubChem CID 71463644
IUPAC Name (1S,2S)-2-N,2-N-bis[(2-diphenylphosphanylphenyl)methyl]cyclohexane-1,2-diamine
SMILES C(N[C@H]1CCCC[C@@H]1NCC1=C(C=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Yellow
Odor Unpleasant
Quantity 100 mg
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 662.78
Percent Purity 97%
Physical Form Solid
Chemical Name or Material (1S,2S)-N,N'-Bis[2-(diphenylphosphino)benzyl]cyclohexane-1,2-diamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'