missing translation for 'onlineSavingsMsg'
Learn More

(1S,3R)-(-)-Camphoric acid, 98%, Thermo Scientific Chemicals

Product Code. 15400478
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 560-09-8
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
MDL Number MFCD00064937
InChI Key LSPHULWDVZXLIL-QUBYGPBYSA-N
Synonym 1s,3r-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid, #NAME?, camphoric acid, camphoric acid,-, 1s,3r-camphoric acid, 1s,3r---camphoric acid, 1s,3r-1,2,2-trimethyl-1,3-cyclopentanedicarboxylic acid, 1,3-cyclopentanedicarboxylic acid, 1,2,2-trimethyl-, 1s,3r, l-camphoric acid, d-+-camphoric acid
PubChem CID 219463
IUPAC Name (1S,3R)-1,2,2-trimethylcyclopentane-1,3-dicarboxylic acid
SMILES CC1(C(CCC1(C)C(=O)O)C(=O)O)C

Specifications

Melting Point 188°C to 190°C
Beilstein 1874674
Merck Index 141733
Formula Weight 200.23
Percent Purity 98%
Quantity 250 mg
Optical Rotation -48° (c=10 in Ethanol)
Chemical Name or Material (1S,3R)-(-)-Camphoric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.