missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% ee, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass Bottle
Specifications
Specifications
Melting Point | 152°C |
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Color | White |
Assay Percent Range | 94% min. (HPLC) |
Infrared Spectrum | Authentic |
Packaging | Glass Bottle |
Solubility Information | Solubility in water: soluble |
Formula Weight | 227.26 |
Percent Purity | 95%,98% ee |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'