missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1S,4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 95%, 98% ee, Thermo Scientific™
£102.54 - £838.00
Chemical Identifiers
CAS | 151907-80-1 |
---|---|
Molecular Formula | C11H17NO4 |
Molecular Weight (g/mol) | 227.26 |
MDL Number | MFCD00211288 |
InChI Key | WOUNTSATDZJBLP-UHFFFAOYNA-N |
Synonym | 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid, 1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid, +-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid, +-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy, cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid |
PubChem CID | 7019281 |
IUPAC Name | (1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid |
SMILES | CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10095073
|
Thermo Scientific Acros
361822500 |
250 mg | Glass Bottle | ||||||
10394252
|
Thermo Scientific
361820010 |
1 g | Glass Bottle |
£269.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10098751
|
Thermo Scientific Acros
361820050 |
5 g | Glass Bottle | ||||||
Description
Chemical Identifiers
151907-80-1 | |
227.26 | |
WOUNTSATDZJBLP-UHFFFAOYNA-N | |
7019281 | |
CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O |
C11H17NO4 | |
MFCD00211288 | |
1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid, 1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid, +-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid, +-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy, cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid | |
(1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid |
Specifications
152°C | |
94.0 | |
White | |
C11H17NO4 | |
MFCD00211288 | |
Solubility in water: soluble | |
CC(C)(C)OC(=O)NC1CC(C=C1)C(O)=O | |
227.26 | |
227.26 | |
Crystalline Powder |
151907-80-1 | |
100.0 | |
94% min. (HPLC) | |
Authentic | |
1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonylamino cyclopent-2-enecarboxylic acid, 1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxylic acid, +-1r,4s-n-boc-4-aminocyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxycarbonyl amino cyclopent-2-ene-1-carboxylic acid, +-1s,4r-n-boc-1-aminocyclopent-2-ene-4-carboxy, cis-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid, 1r,4s-4-tert-butoxy carbonyl amino cyclopent-2-ene-1-carboxylic acid, 1r,4s-rel-4-tert-butoxycarbonyl amino cyclopent-2-enecarboxylic acid | |
WOUNTSATDZJBLP-UHFFFAOYNA-N | |
(1R,4S)-4-[(2-methylpropan-2-yl)oxycarbonylamino]cyclopent-2-ene-1-carboxylic acid | |
7019281 | |
95%,98% ee | |
(1S, 4R)-N-BOC-1-Aminocyclopent-2-ene-4-carboxylic acid, 98% ee |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
RUO – Research Use Only