missing translation for 'onlineSavingsMsg'
Learn More

(1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane, 95%, Thermo Scientific Chemicals

Product Code. 10597054
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113451-59-5
Molecular Formula C10H18N2O2
Molecular Weight (g/mol) 198.26
MDL Number MFCD01569250
InChI Key UXAWXZDXVOYLII-YUMQZZPRSA-N
Synonym 1s,4s-2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-tert-butyl 2,5-diazabicyclo 2.2.1 heptane-2-carboxylate, 1s,4s-n-tert-butoxycarbonyl-2,5-diazabicyclo 2.2.1 heptane, 1s,4s---2-boc-2,5-diazabicyclo 2.2.1 heptane, tert-butyl 1s,4s-2,5-diazabicyclo 2.2.1 heptan-2-carboxylate, 1s,4s-2,5-diaza-bicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, 1s,4s-n-boc-2,5-diazabicyclo 2.2.1 heptane, 1s,4s-2,5-diazabicyclo 2.2.1 heptane-2-carboxylic acid tert-butyl ester, tert-butyl 1s,4s---2,5-diazabicyclo 2.2.1 heptane-2-carboxylate
PubChem CID 11521263
IUPAC Name tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
SMILES CC(C)(C)OC(=O)N1CC2CC1CN2

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 74.0°C to 76.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation -41° to -47° (20°C, 589nm) (C=1, chloroform)
Solubility Information Solubility in water: insoluble
Formula Weight 198.26
Percent Purity 95%
Physical Form Powder
Chemical Name or Material (1S,4S)-(-)-2-BOC-2,5-diazabicyclo[2.2.1]heptane
Show More Show Less
compliance-icons
Product Identifier
  • (1S,4S)-(-)-2-BOC-2,5-diazabicyclo¢2.2.1!heptane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'