Learn More
(1S)-(-)-Camphor, 98%, Thermo Scientific Chemicals
£28.40 - £266.00
Chemical Identifiers
CAS | 464-48-2 |
---|---|
Molecular Formula | C10H16O |
Molecular Weight (g/mol) | 152.24 |
MDL Number | MFCD00064148,MFCD00074738,MFCD00064149,MFCD00064149 |
InChI Key | DSSYKIVIOFKYAU-OMNKOJBGSA-N |
Synonym | --camphor, l-camphor, 1s---camphor, 1s,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, --bornan-2-one, s-camphor, l--camphor, 1s,4s-camphor, camphor,-, --alcanfor |
PubChem CID | 444294 |
ChEBI | CHEBI:15397 |
IUPAC Name | (1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one |
SMILES | CC1(C)C2CC[C@]1(C)C(=O)C2 |
Description
(1S)-(-)-Camphor is used as chiral intermediate and chiral auxiliary precursor. It is used in the synthesis of high-potency sweeteners. It acts as catalytic agent and petrochemical additive.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(1S)-(-)-Camphor is used as chiral intermediate and chiral auxiliary precursor. It is used in the synthesis of high-potency sweeteners. It acts as catalytic agent and petrochemical additive.
Solubility
Soluble in water. (0.34 g/L) at 20°C
Notes
Incompatible materials are oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Chemical Identifiers
464-48-2 | |
152.24 | |
DSSYKIVIOFKYAU-OMNKOJBGSA-N | |
444294 | |
(1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one |
C10H16O | |
MFCD00064148,MFCD00074738,MFCD00064149,MFCD00064149 | |
--camphor, l-camphor, 1s---camphor, 1s,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, --bornan-2-one, s-camphor, l--camphor, 1s,4s-camphor, camphor,-, --alcanfor | |
CHEBI:15397 | |
CC1(C)C2CC[C@]1(C)C(=O)C2 |
Specifications
464-48-2 | |
0.99 | |
65°C (149°F) | |
C10H16O | |
UN2717 | |
14,1732 | |
Soluble in water. (0.34g/L) at 20°C | |
−43° (c=10 in Ethanol) | |
(1S,4S)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-one | |
444294 | |
152.24 | |
(1S)-(-)-Camphor |
175°C to 180°C | |
204°C | |
Camphor-like | |
MFCD00064148,MFCD00074738,MFCD00064149,MFCD00064149 | |
4291747 | |
--camphor, l-camphor, 1s---camphor, 1s,4s-1,7,7-trimethylbicyclo 2.2.1 heptan-2-one, --bornan-2-one, s-camphor, l--camphor, 1s,4s-camphor, camphor,-, --alcanfor | |
DSSYKIVIOFKYAU-OMNKOJBGSA-N | |
CC1(C)C2CC[C@]1(C)C(=O)C2 | |
152.24 | |
CHEBI:15397 | |
98% |
Safety and Handling
GHS H Statement
H228-H315-H319
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P210-P240-P241-P261-P264b-P271-P280i-P281-P302+P352-P304+P340-P305+P351+P338-P308+P313-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335-H340-H500
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: CAMPHOR
missing translation for 'einecsNumber' : 207-354-7
missing translation for 'rtecsNumber' : EX1250000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only