missing translation for 'onlineSavingsMsg'
Learn More

2-(1,2-Benzisoxazol-3-yl)acetic acid, 97%, Thermo Scientific™

Product Code. 10425902
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4865-84-3
Molecular Formula C9H6NO3
Molecular Weight (g/mol) 176.15
MDL Number MFCD02180388
InChI Key BVSIAYQIMUUCRW-UHFFFAOYSA-M
Synonym 2-1,2-benzisoxazol-3-yl acetic acid, 1,2-benzisoxazole-3-acetic acid, 1,2-benzisoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl acetic acid, 2-benzo d isoxazol-3-yl acetic acid, 1,2-benzoxazol-3-ylacetic acid, 2-benzo d isoxazol-3-ylacetic acid, 2-1,2-benzoxazol-3-yl ethanoic acid, nvu, zlchem 388
PubChem CID 301729
SMILES [O-]C(=O)CC1=NOC2=CC=CC=C12

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
IUPAC Name 2-(1,2-benzoxazol-3-yl)acetate
Formula Weight 177.16
Percent Purity 97%
Chemical Name or Material 2-(1, 2-Benzisoxazol-3-yl)acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.