missing translation for 'onlineSavingsMsg'
Learn More

2-(1,3-Benzodioxol-5-yl)ethanol, ≥97%, Thermo Scientific™

Product Code. 10432504
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6006-82-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00666033
InChI Key JADSGOFBFPTCHG-UHFFFAOYSA-N
Synonym 2-benzo d 1,3 dioxol-5-yl ethanol, 2-2h-1,3-benzodioxol-5-yl ethan-1-ol, 3,4-methylenedioxy phenethyl alcohol, 2-1,3-benzodioxol-5-yl ethanol, 1,3-benzodioxole-5-ethanol, homopiperonyl alcohol, 3,4-methylenedioxyphenethyl alcohol, 2-1,3-benzodioxol-5-yl-ethanol, 2-2h-1,3-benzodioxol-5-yl ethanol, benzodioxolylethanol
PubChem CID 2759846
IUPAC Name 2-(1,3-benzodioxol-5-yl)ethanol
SMILES C1OC2=C(O1)C=C(C=C2)CCO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 166.18
Percent Purity 97%
Chemical Name or Material 2-(1, 3-Benzodioxol-5-yl)ethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'