missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazol-4-yl isocyanate, 97%, Thermo Scientific™

Product Code. 10763233
Click to view available options
Quantity:
250 mg
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 342411-14-7
Molecular Formula C7H3N3OS
Molecular Weight (g/mol) 177.181
MDL Number MFCD02681903
InChI Key HPZZFYLZFYDNMA-UHFFFAOYSA-N
PubChem CID 2776284
IUPAC Name 4-isocyanato-2,1,3-benzothiadiazole
SMILES C1=CC2=NSN=C2C(=C1)N=C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Synonym 2,1,3-benzothiadiazol-4-yl isocyanate, 2,1,3-benzothiadiazol-4-yl isoxyanate, 4-isocyanatobenzo c 1,2,5 thiadiazole, 2,1,3-benzothiadiazol-4-ylisocyanate, 2,1,3-benzothiadiazole,4-isocyanato, benzo c 1,2,5-thiadiazol-4-isocyanate, 2,1,3-benzothiadiazole, 4-isocyanato, 2,1,3-benzothiadiazole, 4-isocyanato-9ci
Formula Weight 177.19
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazol-4-yl isocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'