missing translation for 'onlineSavingsMsg'
Learn More

2,1,3-Benzothiadiazole-4-carbaldehyde, 95%, Thermo Scientific™

Product Code. 10732027
5 g, 5g
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10732027

Brand: Thermo Scientific Maybridge CC09104DE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5170-68-3
Molecular Formula C7H4N2OS
Molecular Weight (g/mol) 164.182
MDL Number MFCD02681902
InChI Key ANVJARPTPIVPRC-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carboxaldehyde, tos-bb-1233, 2,1,3-benzothiadiazol-4-aldehyde, 2,1,3-benzothiadiazole-4-aldehyde, 2,1,3-benzothiadiazol-4-carboxaldehyde, benzo c 1,2,5-thiadiazole-4-carbaldehyde, 2,1,3-benzothiadiazole-4-carbaldehydeShow MoreShow Less
PubChem CID 2776283
IUPAC Name 2,1,3-benzothiadiazole-4-carbaldehyde
SMILES C1=CC2=NSN=C2C(=C1)C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 164.19
Percent Purity 95%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-4-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,1,3-Benzothiadiazole-4-carbaldehyde, 95%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.