missing translation for 'onlineSavingsMsg'
Learn More
2,1,3-Benzothiadiazole-5-carbaldehyde, 97%, Thermo Scientific™ Product Code.: 10262632

2,1,3-Benzothiadiazole-5-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10262632
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 10262632

Brand: Thermo Scientific Maybridge CC09204CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 71605-72-6
Molecular Formula C7H4N2OS
Molecular Weight (g/mol) 164.182
MDL Number MFCD00457949
InChI Key GEFIFDVQYCPLHC-UHFFFAOYSA-N
Synonym benzo c 1,2,5 thiadiazole-5-carbaldehyde, 2,1,3-benzothiadiazole-5-carboxaldehyde, benzo 1,2,5 thiadiazole-5-carbaldehyde, 5-formyl-2,1,3-benzothiadiazole, 5-formylbenzo-2,1,3-thiadiazole, benzo 1,2,5 thiadiazol-5-carbaldehyde, benzo 1,2,5 thiadiazole-5-carboxaldehyde
PubChem CID 2776290
IUPAC Name 2,1,3-benzothiadiazole-5-carbaldehyde
SMILES C1=CC2=NSN=C2C=C1C=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 164.19
Percent Purity 97%
Chemical Name or Material 2, 1, 3-Benzothiadiazole-5-carbaldehyde
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,1,3-Benzothiadiazole-5-carbaldehyde, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.