missing translation for 'onlineSavingsMsg'
Learn More

2-(1,4-Diazepan-1-yl)nicotinonitrile, ≥95%, Thermo Scientific™

Product Code. 10561112
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10561112

Brand: Thermo Scientific Maybridge MO00102DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 352018-97-4
Molecular Formula C11H14N4
Molecular Weight (g/mol) 202.261
MDL Number MFCD03086210
InChI Key KQOHMUWBQXSUAK-UHFFFAOYSA-N
Synonym 2-1,4-diazepan-1-yl nicotinonitrile, 2-1,4-diazepan-1-yl pyridine-3-carbonitrile, 3-pyridinecarbonitrile,2-hexahydro-1h-1,4-diazepin-1-yl, 2-homopiperazin-1-yl nicotinonitrile, 2-1,4 diazepan-1-yl-nicotinonitrile, 2-1,4-diazaperhydroepinyl pyridine-3-carbonitrile
PubChem CID 2779669
IUPAC Name 2-(1,4-diazepan-1-yl)pyridine-3-carbonitrile
SMILES C1CNCCN(C1)C2=C(C=CC=N2)C#N

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 202.26
Percent Purity 95%
Chemical Name or Material 2-(1, 4-Diazepan-1-yl)nicotinonitrile
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.