missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Adamantyl)acetic acid, 97%, Thermo Scientific™

Product Code. 10567074
Click to view available options
Quantity:
1 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 4942-47-6
Molecular Formula C12H17O2
Molecular Weight (g/mol) 193.27
MDL Number MFCD00074728
InChI Key AOTQGWFNFTVXNQ-UHFFFAOYSA-M
Synonym 1-adamantaneacetic acid, 2-adamantan-1-yl acetic acid, 1-adamantylacetic acid, 1-adamantane acetic acid, adamantan-1-ylacetic acid, 2-1-adamantyl acetic acid, tricyclo 3.3.1.13,7 dec-1-ylacetic acid, adamantan-1-yl-acetic acid, 1-carboxymethyl adamantane
PubChem CID 123221
SMILES [O-]C(=O)CC12CC3CC(CC(C3)C1)C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name 2-(adamantan-1-yl)acetate
Formula Weight 194.27
Percent Purity 97%
Chemical Name or Material 2-(1-Adamantyl)acetic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.