missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Benzy-4-piperidinyloxy)-N,N-dimethylacetamide, Thermo Scientific™

Product Code. 11484890
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902836-18-4
Molecular Formula C16H24N2O2
Molecular Weight (g/mol) 276.38
MDL Number MFCD08061032
InChI Key WYGXZOIPPPSCFH-UHFFFAOYSA-N
Synonym 2-1-benzylpiperidin-4-yloxy-n,n-dimethylacetamide, 2-1-benzy-4-piperidinyloxy-n,n-dimethylacetamide, n,n-dimethyl-2-1-benzyl 4-piperidyloxy acetamide, 2-1-benzylpiperidin-4-yl oxy-n,n-dimethylacetamide
PubChem CID 24208843
IUPAC Name 2-(1-benzylpiperidin-4-yl)oxy-N,N-dimethylacetamide
SMILES CN(C)C(=O)COC1CCN(CC2=CC=CC=C2)CC1

Specifications

Quantity 1 g
Formula Weight 276.38
Chemical Name or Material 2-(1-Benzy-4-piperidinyloxy)-N,N-dimethylacetamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'