missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Methylhydrazino)-4-(trifluoromethyl)pyrimidine, 97%, Thermo Scientific™

Product Code. 15458555
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1372711-01-7
Molecular Formula C6H7F3N4
Molecular Weight (g/mol) 192.145
MDL Number MFCD20037418
InChI Key KSHPFMMNRGWDRW-UHFFFAOYSA-N
Synonym 2-1-methylhydrazin-1-yl-4-trifluoromethyl pyrimidine, 2-1-methylhydrazino-4-trifluoromethyl pyrimidine, 2-1-methylhydrazino-4-trifluoromethyl-pyrimidine
PubChem CID 73995159
IUPAC Name 1-methyl-1-[4-(trifluoromethyl)pyrimidin-2-yl]hydrazine
SMILES CN(C1=NC=CC(=N1)C(F)(F)F)N

Specifications

Quantity 1 g
Formula Weight 192.14
Percent Purity 97%
Chemical Name or Material 2-(1-Methylhydrazino)-4-(trifluoromethyl)pyrimidine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'