missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1-Naphthyl)ethanol, 95%, Thermo Scientific™
£53.90 - £218.80
Chemical Identifiers
CAS | 773-99-9 |
---|---|
Molecular Formula | C12H12O |
Molecular Weight (g/mol) | 172.23 |
InChI Key | RXWNCMHRJCOWDK-UHFFFAOYSA-N |
Synonym | 1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol |
PubChem CID | 13047 |
IUPAC Name | 2-naphthalen-1-ylethanol |
SMILES | C1=CC=C2C(=C1)C=CC=C2CCO |
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
773-99-9 | |
172.23 | |
1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol | |
2-naphthalen-1-ylethanol |
C12H12O | |
RXWNCMHRJCOWDK-UHFFFAOYSA-N | |
13047 | |
C1=CC=C2C(=C1)C=CC=C2CCO |
Specifications
773-99-9 | |
100.0 | |
White | |
96°C | |
94% min. (HPLC) | |
C12H12O | |
06,668 | |
RXWNCMHRJCOWDK-UHFFFAOYSA-N | |
2-naphthalen-1-ylethanol | |
13047 | |
95% | |
2-(1-Naphthyl)ethanol |
94.0 | |
61.0°C to 65.0°C | |
186°C (23.0 hPa) | |
Authentic | |
Glass bottle | |
C10H7CH2CH2OH | |
1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol | |
C1=CC=C2C(=C1)C=CC=C2CCO | |
172.23 | |
172.23 | |
Crystalline Powder or Solid |
Safety and Handling
missing translation for 'einecsNumber' : 212-260-4