missing translation for 'onlineSavingsMsg'
Learn More

2-(1-Naphthyl)ethanol, 95%, Thermo Scientific™

Product Code. 11944651
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 773-99-9
Molecular Formula C12H12O
Molecular Weight (g/mol) 172.23
InChI Key RXWNCMHRJCOWDK-UHFFFAOYSA-N
Synonym 1-naphthaleneethanol, 2-1-naphthyl ethanol, 2-naphthalen-1-yl ethanol, 1-2-hydroxyethyl naphthalene, naphthalen-1-ethanol, 2-naphthylethanol, 1-naphthyl ethanol, 1-naphthalene ethanol, 2-naphthalen-1-yl ethan-1-ol, 1-napthaleneethanol
PubChem CID 13047
IUPAC Name 2-naphthalen-1-ylethanol
SMILES C1=CC=C2C(=C1)C=CC=C2CCO

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 61.0°C to 65.0°C
Color White
Boiling Point 186°C (23.0 hPa)
Flash Point 96°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula C10H7CH2CH2OH
Quantity 50 g
Beilstein 06,668
Formula Weight 172.23
Percent Purity 95%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 2-(1-Naphthyl)ethanol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'