missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1-Pyrrolidinyl)nicotinic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
5 g
Specifications
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Quantity | 5 g |
IUPAC Name | 2-pyrrolidin-1-ylpyridine-3-carboxylic acid |
Formula Weight | 192.22 |
Percent Purity | 95% |
Chemical Name or Material | 2-(1-Pyrrolidinyl)nicotinic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction