missing translation for 'onlineSavingsMsg'
Learn More
2-[(11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate, 97+%, Thermo Scientific™ Product Code.: 15425455

2-[(11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate, 97+%, Thermo Scientific™

Product Code. 15425455
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15425455

Brand: Thermo Scientific Alfa Aesar H60215.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1223007-01-9
Molecular Formula C24H23BF4NP
Molecular Weight (g/mol) 443.232
MDL Number MFCD17018773
InChI Key AMEHGPFCNMMXTB-UHFFFAOYSA-O
Synonym 2-11br-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11bs-3h-dinaphtho 2,1-c:1',2'-e phosphepin-4 5h-yl ethylammonium tetrafluoroborate, 2-11br-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-11bs-3,5-dihydro-4h-dinaphtho 2,1-c:1',2'-e phosphepin-4-yl ethaminium tetrafluorborate, 2-13-phosphapentacyclo 13.8.0.0_,__.0_,?.0_?,__ tricosa-1 15 ,2,4,6,8,10,16,18,20,22-decaen-13-yl ethanaminium tetrafluoroborateShow MoreShow Less
PubChem CID 72376450
SMILES [B-](F)(F)(F)F.C1C2=C(C3=CC=CC=C3C=C2)C4=C(CP1CC[NH3+])C=CC5=CC=CC=C54

Specifications

Quantity 250 mg
Formula Weight 443.22
Percent Purity ≥97%
Chemical Name or Material 2-[(11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-[(11bR)-3H-Dinaphtho[2,1-c:1',2'-e]phosphepin-4(5H)-yl]ethylammonium tetrafluoroborate, 97+%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.