missing translation for 'onlineSavingsMsg'
Learn More

2-(1H-1,2,4-triazol-1-ylmethyl)aniline, 97%, Thermo Scientific™

Product Code. 10133474
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 127988-21-0
Molecular Formula C9H10N4
Molecular Weight (g/mol) 174.207
MDL Number MFCD09025845
InChI Key CBGQHZNXHUCAGE-UHFFFAOYSA-N
Synonym 2-1h-1,2,4-triazol-1-ylmethyl aniline, 2-1,2,4-triazol-1-yl methyl aniline, 2-1,2,4-triazol-1-ylmethyl aniline, 2-1h-1,2,4-triazol-1-ylmethyl benzeneamine, benzenamine,2-1h-1,2,4-triazol-1-ylmethyl, acmc-20msof, 2-1,2,4-triazolylmethyl phenylamine
PubChem CID 6482003
IUPAC Name 2-(1,2,4-triazol-1-ylmethyl)aniline
SMILES C1=CC=C(C(=C1)CN2C=NC=N2)N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 174.21
Percent Purity 97%
Chemical Name or Material 2-(1H-1, 2, 4-Triazol-1-ylmethyl)aniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'