missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(1H-Pyrrol-1-yl)benzoic acid, ≥97%, Thermo Scientific™
Click to view available options
Amber Glass Bottle; 1g
Specifications
CAS | 10333-68-3 |
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Molecular Formula | C11H9NO2 |
MDL Number | MFCD00051645 |
Quantity | 1g |
Synonym | 2-1h-pyrrol-1-yl benzoic acid, 2-1-pyrrolyl benzoic acid, 1-2-carboxyphenyl pyrrole, 2-pyrrol-1-yl benzoic acid, 2-pyrrol-1-yl-benzoic acid, 2-pyrrolylbenzoic acid, maybridge1_002677, 1-o-carboxyphenyl pyrrole, n-2-carboxyphenyl pyrrole, 1h-pyrrol-1-ylbenzoic acid |
InChI Key | GNWTWXOZRSBCOZ-UHFFFAOYSA-N |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'