Learn More
2,2,2-Trifluoroacetamide, 97%, Thermo Scientific Chemicals
£26.90 - £282.00
Chemical Identifiers
CAS | 354-38-1 |
---|---|
Molecular Formula | C2H2F3NO |
Molecular Weight (g/mol) | 113.04 |
MDL Number | MFCD00008008 |
InChI Key | NRKYWOKHZRQRJR-UHFFFAOYSA-N |
Synonym | trifluoroacetamide, acetamide, 2,2,2-trifluoro, unii-36mu16byfk, n-trifluoroacetamide, 2,2,2-trifluoro-acetamide, 36mu16byfk, trifluroacetamide, trifluoro-acetamide, snylgqhjptadtaetp@, pubchem12622 |
PubChem CID | 67717 |
IUPAC Name | 2,2,2-trifluoroacetamide |
SMILES | NC(=O)C(F)(F)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11407117
|
Thermo Scientific Alfa Aesar
A14451.14 |
25 g |
£26.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11417117
|
Thermo Scientific Alfa Aesar
A14451.22 |
100 g |
£85.50
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11427117
|
Thermo Scientific Alfa Aesar
A14451.36 |
500 g |
£282.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2,2,2-Trifluoroacetamide used in a convenient alternative to the Gabriel synthesis of primary amines from halides by N-alkylation followed by cleavage of the readily-hydrolyzed trifluoroacetyl group and for improved N-alkylation under phase-transfer conditions allowing successive alkylation with different alkyl groups.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,2,2-Trifluoroacetamide used in a convenient alternative to the Gabriel synthesis of primary amines from halides by N-alkylation followed by cleavage of the readily-hydrolyzed trifluoroacetyl group and for improved N-alkylation under phase-transfer conditions allowing successive alkylation with different alkyl groups.
Solubility
Soluble in water (460 g/L at 20°C), alcohol, ether, chloroform (partially), and methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
Chemical Identifiers
354-38-1 | |
113.04 | |
NRKYWOKHZRQRJR-UHFFFAOYSA-N | |
67717 | |
NC(=O)C(F)(F)F |
C2H2F3NO | |
MFCD00008008 | |
trifluoroacetamide, acetamide, 2,2,2-trifluoro, unii-36mu16byfk, n-trifluoroacetamide, 2,2,2-trifluoro-acetamide, 36mu16byfk, trifluroacetamide, trifluoro-acetamide, snylgqhjptadtaetp@, pubchem12622 | |
2,2,2-trifluoroacetamide |
Specifications
354-38-1 | |
162°C to 164°C | |
MFCD00008008 | |
trifluoroacetamide, acetamide, 2,2,2-trifluoro, unii-36mu16byfk, n-trifluoroacetamide, 2,2,2-trifluoro-acetamide, 36mu16byfk, trifluroacetamide, trifluoro-acetamide, snylgqhjptadtaetp@, pubchem12622 | |
NRKYWOKHZRQRJR-UHFFFAOYSA-N | |
2,2,2-trifluoroacetamide | |
67717 | |
97% |
70°C to 75°C | |
C2H2F3NO | |
1753625 | |
Soluble in water (460g/L at 20°C),alcohol,ether,chloroform (partially),and methanol. | |
NC(=O)C(F)(F)F | |
113.04 | |
113.04 | |
2,2,2-Trifluoroacetamide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 206-559-9
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only