missing translation for 'onlineSavingsMsg'
Learn More

2,2,2-Trimethylthioacetamide, 97%, Thermo Scientific Chemicals

Product Code. 11360036
Click to view available options
Quantity:
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2,2-Trimethylthioacetamide, is used as a pharmaceutical and organic intermediate, an important amino protective agent, pharmaceutical and synthetic organic make intermediates, organic synthesis used to introduce t- Boc protect gene species.

Solubility
Soluble in water (10 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-22-8
Molecular Formula C5H11NS
Molecular Weight (g/mol) 117.21
MDL Number MFCD09742834
InChI Key FJZJUSOFGBXHCV-UHFFFAOYSA-N
Synonym 2,2,2-trimethylthioacetamide, 2,2-dimethylthiopropionamide, neopentanethioamide, propanethioamide, 2,2-dimethyl, thiopivalamide, 2,2-dimethyl-thiopropionamide, 2,2-dimethylthio propionamide, propanethioamide,2,2-dimethyl, 2-methyl-2-methylthiopropionamide, propionamide, 2-methyl-2-methylthio
PubChem CID 3031130
IUPAC Name 2,2-dimethylpropanethioamide
SMILES CC(C)(C)C(=S)N

Specifications

Melting Point 113°C to 117°C
Quantity 10 g
Solubility Information Soluble in water (10g/L) (25°C).
Formula Weight 117.21
Percent Purity 97%
Chemical Name or Material 2,2,2-Trimethylthioacetamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'