Learn More
2,2,3,3,3-Pentafluoropropionamide, 97%, Thermo Scientific™
£28.10 - £97.40
Chemical Identifiers
CAS | 354-76-7 |
---|---|
Molecular Formula | C3H2F5NO |
Molecular Weight (g/mol) | 163.05 |
MDL Number | MFCD00039771 |
InChI Key | KQTOYEUYHXUEDB-UHFFFAOYSA-N |
PubChem CID | 67722 |
IUPAC Name | pentafluoropropanamide |
SMILES | NC(=O)C(F)(F)C(F)(F)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2,2,3,3,3-Pentafluoropropionamide, is used as a chemical, organic, pharamceutical intermediate.
Solubility
Soluble in methanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
Chemical Identifiers
354-76-7 | |
163.05 | |
KQTOYEUYHXUEDB-UHFFFAOYSA-N | |
pentafluoropropanamide |
C3H2F5NO | |
MFCD00039771 | |
67722 | |
NC(=O)C(F)(F)C(F)(F)F |
Specifications
354-76-7 | |
C3H2F5NO | |
1773386 | |
Soluble in methanol. | |
NC(=O)C(F)(F)C(F)(F)F | |
163.05 | |
163.05 | |
2,2,3,3,3-Pentafluoropropionamide |
92°C to 97°C | |
MFCD00039771 | |
pentafluoropropionamide, pentafluoropropanamide, propanamide, 2,2,3,3,3-pentafluoro, 2,2,3,3,3-pentafluoropropionamide, acmc-1cjql, pentafluoro propionic acid amide, 2,2,3,3,3-pentafluoropropionamide,, propanamide,2,2,3,3,3-pentafluoro | |
KQTOYEUYHXUEDB-UHFFFAOYSA-N | |
pentafluoropropanamide | |
67722 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 206-569-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures