missing translation for 'onlineSavingsMsg'
Learn More

2,2,3,3,3-Pentafluoropropylamine, 97%, Thermo Scientific Chemicals

Product Code. 11363766
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-03-7
Molecular Formula C3H4F5N
Molecular Weight (g/mol) 149.064
MDL Number MFCD00042459
InChI Key DPQNQLKPUVWGHE-UHFFFAOYSA-N
PubChem CID 79002
IUPAC Name 2,2,3,3,3-pentafluoropropan-1-amine
SMILES C(C(C(F)(F)F)(F)F)N

Specifications

Density 1.4
Boiling Point 49°C to 50°C
Flash Point >100°C (212°F)
Refractive Index 1.297
Quantity 1 g
UN Number UN2735
Beilstein 1743135
Synonym 2,2,3,3,3-pentafluoropropylamine, pentafluoropropylamine, acmc-1amc1, 1h,1h-pentafluoropropylamine, 2,2,3,3,3-penta-fluoropropylamine, 2,2,3,3,3-pentafluoro-propylamine, 2,2,3,3,3-pentafluoro-propyl amine, 1-propanamine,2,2,3,3,3-pentafluoro, 1-propanamine, 2,2,3,3,3-pentafluoro
Formula Weight 149.06
Percent Purity 97%
Chemical Name or Material 2,2,3,3,3-Pentafluoropropylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'