missing translation for 'onlineSavingsMsg'
Learn More

2-(2,3-Dihydro-1-benzofuran-5-yl)-4-methyl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10291133
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 690632-04-3
Molecular Formula C13H11NO3S
Molecular Weight (g/mol) 261.295
MDL Number MFCD01313483
InChI Key DCXMMNGTUXDRND-UHFFFAOYSA-N
Synonym 2-2,3-dihydro-1-benzofuran-5-yl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-2,3-dihydrobenzofuran-5-yl-4-methylthiazole-5-carboxylic acid, 2-2,3-dihydro-1-benzo b furan-5-yl-4-methyl-1,3-thiazole-5-carboxylic acid, 2-2,3-dihydrobenzo b furan-5-yl-4-methyl-1,3-thiazole-5-carboxylic acid
PubChem CID 2798545
IUPAC Name 2-(2,3-dihydro-1-benzofuran-5-yl)-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC1=C(SC(=N1)C2=CC3=C(C=C2)OCC3)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 261.3
Percent Purity 97%
Chemical Name or Material 2-(2, 3-Dihydro-1-benzofuran-5-yl)-4-methyl-1, 3-thiazole-5-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'