missing translation for 'onlineSavingsMsg'
Learn More

2,2',4,4'-Tetrahydroxybenzophenone, 98+%

Product Code. 11403943
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11403943

Brand: Thermo Scientific Alfa Aesar A10237.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 131-55-5 | C13H10O5 | 246.218 g/mol

2,2',4,4'-Tetrahydroxybenzophenone is used as the raw material of ultraviolet absorbers in cosmetics, plastics, resins and rubber. It is used as pharmaceutical intermediates and light-sensitive materials. It is also used as pharmaceutical adjuvant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′,4,4′-Tetrahydroxybenzophenone is used as the raw material of ultraviolet absorbers in cosmetics, plastics, resins and rubber. It is used as pharmaceutical intermediates and light-sensitive materials. It is also used as pharmaceutical adjuvant.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131-55-5
Molecular Formula C13H10O5
Molecular Weight (g/mol) 246.218
MDL Number MFCD00002278
InChI Key WXNRYSGJLQFHBR-UHFFFAOYSA-N
Synonym 2,2',4,4'-tetrahydroxybenzophenone, bis 2,4-dihydroxyphenyl methanone, benzophenone-2, uvinol d-50, uvinul d-50, methanone, bis 2,4-dihydroxyphenyl, 2,4,2',4'-tetrahydroxybenzophenone, unii-prr8k3h9vn, 2,2',4,4'-tetrehydroxybenzophenone, prr8k3h9vn
PubChem CID 8571
IUPAC Name bis(2,4-dihydroxyphenyl)methanone
SMILES C1=CC(=C(C=C1O)O)C(=O)C2=C(C=C(C=C2)O)O

Specifications

Melting Point 198°C to 203°C
Density 1.216
Quantity 100 g
Beilstein 1914746
Solubility Information Slightly soluble in water.
Formula Weight 246.22
Percent Purity ≥98%
Chemical Name or Material 2,2',4,4'-Tetrahydroxybenzophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.