missing translation for 'onlineSavingsMsg'
Learn More

2-(2,4-Dichlorophenoxy)aniline hydrochloride, 98%, Thermo Scientific™

Product Code. 11484227
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89279-16-3
Molecular Formula C12H10Cl3NO
Molecular Weight (g/mol) 290.568
MDL Number MFCD00052001
InChI Key VPOKWNPEXKYIPO-UHFFFAOYSA-N
Synonym 2-2,4-dichlorophenoxy aniline hydrochloride, benzenamine, 4-2,4-dichlorophenoxy-, hydrochloride, 4-2,4-dichlorophenoxy benzenamine hydrochloride, 2-2,4-dichlorophenoxy aniline hcl, 2-2,4-dichloro-phenoxy aniline hcl, 2-2,4-dichloro-phenoxy aniline hydrochloride
PubChem CID 2795482
IUPAC Name 2-(2,4-dichlorophenoxy)aniline;hydrochloride
SMILES C1=CC=C(C(=C1)N)OC2=C(C=C(C=C2)Cl)Cl.Cl

Specifications

Melting Point 96°C to 98°C
Quantity 5 g
Beilstein 3736700
Formula Weight 290.58
Percent Purity 98%
Chemical Name or Material 2-(2,4-Dichlorophenoxy)aniline hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.