missing translation for 'onlineSavingsMsg'
Learn More
2,2,4-Trimethyl-1,3-dioxolane, 99%, Thermo Scientific Chemicals Product Code.: 11434857

2,2,4-Trimethyl-1,3-dioxolane, 99%, Thermo Scientific Chemicals

Product Code. 11434857
25 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11434857

Brand: Thermo Scientific Alfa Aesar B21823.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-11-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00090841
InChI Key ALTFLAPROMVXNX-UHFFFAOYNA-N
PubChem CID 62384
IUPAC Name 2,2,4-trimethyl-1,3-dioxolane
SMILES CC1COC(C)(C)O1

Specifications

Density 0.897
Boiling Point 98°C
Flash Point 32°C (90°F)
Quantity 25 g
UN Number UN3271
Synonym propylene glycol acetone ketal, 1,3-dioxolane, 2,2,4-trimethyl, acetone propylene glycol acetal, 1,3-dioxolane,2,2,4-trimethyl, fema no. 3441, 2,2,4-trimethyl-1,3-oxacyclopentane, 2,2,4-trimethyl-1,3-dioxacyclopentane, propylene glycol acetonide, 1,2-propanediol acetonide, 2,2,4-trimethyldioxolane
Formula Weight 116.16
Percent Purity 99%
Chemical Name or Material 2,2,4-Trimethyl-1,3-dioxolane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10216533 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog NumberB21823.36,B21823.14,B21823.22
Lot Number10250934 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberB21823.14,B21823.36,B21823.22
Lot Number10248579 Certificate TypeCertificate of Analysis Date06/11/2023 Catalog NumberB21823.14,B21823.36,B21823.22
Lot Number10246984 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberB21823.14,B21823.36,B21823.22
Lot Number10242585 Certificate TypeCertificate of Analysis Date04/08/2022 Catalog NumberB21823.14,B21823.36,B21823.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2,4-Trimethyl-1,3-dioxolane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.