missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethyl-1,3-pentanediol diisobutyrate, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
1 kg
Packaging:
Glass bottle
Unit Size:
1kg
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
| CAS Min % | 97.5 |
| CAS Max % | 100.0 |
| Melting Point | -70.0°C |
| Color | Colorless |
| Density | 0.9400g/mL |
| Boiling Point | 280.0°C |
| Flash Point | 128°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 98% |
| Packaging | Glass bottle |
| Afficher plus |
Sécurité et traitement
Identifiant produit
- 2,2,4-Trimethyl-1,3-pentanediol diisobutyrate
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu