missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2,4-Trimethylpentane, ≥99.5% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -107°C |
Color | Colorless |
Linear Formula | (CH3)2CHCH2C(CH3)3 |
UN Number | UN1262 |
Beilstein | 1696876 |
Formula Weight | 114.23g/mol |
Percent Purity | ≥99.5% (GC) |
Boiling Point | 98°C to 100°C |
Quantity | 2.5 L |
Explosion Limit | 6% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'