missing translation for 'onlineSavingsMsg'
Learn More

2-(2,5-Dimethyl-1H-pyrrol-1-yl)benzoic acid, 97%, Thermo Scientific™

Product Code. 10780924
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 92028-57-4
Molecular Formula C13H13NO2
Molecular Weight (g/mol) 215.25
MDL Number MFCD01125264
InChI Key ZLYUUANOICYAAL-UHFFFAOYSA-N
Synonym 2-2,5-dimethyl-1h-pyrrol-1-yl benzoic acid, 2-2,5-dimethylpyrrol-1-yl benzoic acid, 2-2,5-dimethyl-1-pyrrolyl benzoic acid, 2-2,5-dimethyl-pyrrol-1-yl-benzoic acid, 2-2,5-dimethylpyrrolyl benzoic acid, cbmicro_048042, benzoic acid,2-2,5-dimethyl-1h-pyrrol-1-yl
PubChem CID 292956
IUPAC Name 2-(2,5-dimethylpyrrol-1-yl)benzoic acid
SMILES CC1=CC=C(C)N1C1=CC=CC=C1C(O)=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 215.25
Percent Purity 97%
Chemical Name or Material 2-(2, 5-Dimethyl-1H-pyrrol-1-yl)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'