missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Used as reactant for synthesis of Bis-cycloiridiated complexes for electroluminescent studies, cationic imido alkylidene complexes, polyaromatic ligands containing pyrene and mono-cyclometalated platinum complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 127°C |
Boiling Point | 180°C (0.05 mmHg) |
Quantity | 1 g |
Solubility Information | Soluble in acetone and methanol. |
Formula Weight | 190.24 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | (2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'