Learn More
2,2,6,6-Tetramethylpiperidine, 98%, Thermo Scientific Chemicals
£44.72 - £225.12
Chemical Identifiers
CAS | 768-66-1 |
---|---|
Molecular Formula | C9H19N |
Molecular Weight (g/mol) | 141.26 |
InChI Key | RKMGAJGJIURJSJ-UHFFFAOYSA-N |
Synonym | norpempidine, piperidine, 2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl piperidine, unii-44n9s1ycfm, 2,2,6,6-tetramethylpeperidine, 44n9s1ycfm, 2,2,6,6 tetramethyl piperidine, 2,2,6,6-tetramethyl-piperidine, pubchem7259, acmc-209p6i |
PubChem CID | 13035 |
IUPAC Name | 2,2,6,6-tetramethylpiperidine |
SMILES | CC1(CCCC(N1)(C)C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10561591
|
Thermo Scientific Acros
151950100 |
10 g | Glass bottle |
£44.72
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10436800
|
Thermo Scientific Acros
151950250 |
25 g | Glass bottle |
£85.43
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11926301
|
Thermo Scientific Acros
151951000 |
100 g | Glass bottle |
£225.12
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
768-66-1 | |
141.26 | |
norpempidine, piperidine, 2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl piperidine, unii-44n9s1ycfm, 2,2,6,6-tetramethylpeperidine, 44n9s1ycfm, 2,2,6,6 tetramethyl piperidine, 2,2,6,6-tetramethyl-piperidine, pubchem7259, acmc-209p6i | |
2,2,6,6-tetramethylpiperidine |
C9H19N | |
RKMGAJGJIURJSJ-UHFFFAOYSA-N | |
13035 | |
CC1(CCCC(N1)(C)C)C |
Specifications
768-66-1 | |
0.8300g/mL | |
24°C | |
97.5% min. (GC) | |
1.4430 to 1.4460 | |
0.83 | |
Solubility in water: miscible. Other solubilities: soluble in toluene, dimethylformamide | |
CC1(CCCC(N1)(C)C)C | |
141.26 | |
141.26 | |
Liquid |
Colorless to Yellow | |
152.0°C | |
Authentic | |
C9H19N | |
20,129 | |
norpempidine, piperidine, 2,2,6,6-tetramethyl, 2,2,6,6-tetramethyl piperidine, unii-44n9s1ycfm, 2,2,6,6-tetramethylpeperidine, 44n9s1ycfm, 2,2,6,6 tetramethyl piperidine, 2,2,6,6-tetramethyl-piperidine, pubchem7259, acmc-209p6i | |
RKMGAJGJIURJSJ-UHFFFAOYSA-N | |
2,2,6,6-tetramethylpiperidine | |
13035 | |
98% | |
2,2,6,6-Tetramethylpiperidine |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective glo
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-199-3
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 C
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only