missing translation for 'onlineSavingsMsg'
Learn More

2-(2-Aminoethoxy)ethanol, 98%, Thermo Scientific Chemicals

Product Code. 10519630
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 929-06-6
Molecular Formula C4H11NO2
Molecular Weight (g/mol) 105.14
InChI Key GIAFURWZWWWBQT-UHFFFAOYSA-N
Synonym 2-2-aminoethoxy ethanol, diglycolamine, 2-aminoethoxyethanol, ethanol, 2-2-aminoethoxy, diethylene glycol amine, 2-2-hydroxyethoxy ethylamine, diethylene glycol monoamine, 2-amino-2'-hydroxydiethyl ether, 1-amino-2-2-hydroxyethoxy ethane, 5-hydroxy-3-oxapentylamine
PubChem CID 13578
IUPAC Name 2-(2-aminoethoxy)ethanol
SMILES C(COCCO)N

Specifications

Melting Point -12.5°C
Color Yellow
Density 1.0600g/mL
Boiling Point 218.0°C to 224.0°C
Flash Point 127°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula H2NCH2CH2OCH2CH2OH
Refractive Index 1.4585 to 1.4605
Quantity 1 L
Beilstein 04,III,642
Specific Gravity 1.06
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol and aromatic hydrocarbons, practically immiscible with aliphatic hydrocarbons, and ethyl ether
Formula Weight 105.14
Percent Purity 98%
Physical Form Crystalline Powder and/or Chunks
Chemical Name or Material 2-(2-Aminoethoxy)ethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2-(2-Aminoethoxy)ethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'