Learn More
2-(2-Aminoethyl)pyridine, 95%, Thermo Scientific Chemicals
£21.90 - £400.00
Chemical Identifiers
CAS | 2706-56-1 |
---|---|
Molecular Formula | C7H10N2 |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00006367 |
InChI Key | XPQIPUZPSLAZDV-UHFFFAOYSA-N |
Synonym | 2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine |
PubChem CID | 75919 |
ChEBI | CHEBI:74024 |
IUPAC Name | 2-pyridin-2-ylethanamine |
SMILES | NCCC1=CC=CC=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10508523
|
Thermo Scientific Acros
103690010 |
1 g | Glass bottle |
£21.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10211473
|
Thermo Scientific Acros
103690100 |
10 g | Glass bottle |
£108.00
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10517373
|
Thermo Scientific Acros
103690500 |
50 g | Glass bottle |
£400.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
2706-56-1 | |
122.17 | |
XPQIPUZPSLAZDV-UHFFFAOYSA-N | |
75919 | |
2-pyridin-2-ylethanamine |
C7H10N2 | |
MFCD00006367 | |
2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine | |
CHEBI:74024 | |
NCCC1=CC=CC=N1 |
Specifications
2706-56-1 | |
100.0 | |
1.0210g/mL | |
100°C | |
94% min. (GC) | |
(C5H4N)CH2CH2NH2 | |
MFCD00006367 | |
1.021 | |
Solubility in water: soluble. Other solubilities: soluble in most common polar solvents,insoluble in non-polar solvents and oils | |
NCCC1=CC=CC=N1 | |
122.17 | |
CHEBI:74024 | |
95% | |
2-(2-Aminoethyl)pyridine |
94.0 | |
Colorless to Yellow | |
105°C to 106°C (10.0 mmHg) | |
Authentic | |
C7H10N2 | |
1.5340 to 1.538 | |
22, 434 | |
2-2-aminoethyl pyridine, 2-pyridylethylamine, 2-2-pyridyl ethylamine, 2-pyridin-2-yl ethanamine, 2-pyridineethanamine, demethylbetahistine, 2-pyridin-2-yl ethan-1-amine, alpha-pyridylethylamine, pyridine, 2-2-aminoethyl, 2-aminoethylpyridine | |
XPQIPUZPSLAZDV-UHFFFAOYSA-N | |
2-pyridin-2-ylethanamine | |
75919 | |
122.17 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-295-1
RUO – Research Use Only