missing translation for 'onlineSavingsMsg'
Learn More
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%, Thermo Scientific Chemicals Product Code.: 15460447

2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%, Thermo Scientific Chemicals

Product Code. 15460447
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15460447

Brand: Thermo Scientific Alfa Aesar J65535.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chromogenic substrate for peroxidase assays

Chromogenic substrate for peroxidase assays.2,2'-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chromogenic substrate for peroxidase assays.2,2′-Azino-bis(3-ethylbenzthiazoline-6-sulfonic acid) diammonium salt is widely utilized as a peroxidase substrate suitable for use in enzyme-linked immunosorbent assay (ELISA) procedures. It plays an important role in the food industry to measure the antioxidant capacities of foods. It is used to observe the reaction kinetics of specific enzymes and involved in glucose estimating reagent in blood serum.

Solubility
Dissolve in water at 50mg/ml. Solution may be slightly hazy.

Notes
Light Sensitive. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30931-67-0
Molecular Formula C18H24N6O6S4
Molecular Weight (g/mol) 548.67
MDL Number MFCD00010404,MFCD00010404
InChI Key OHDRQQURAXLVGJ-AXMZSLBLSA-N
Synonym ammonium 2,2'-hydrazine-1,2-diylidene bis 3-ethyl-2,3-dihydrobenzo d thiazole-6-sulfonate
PubChem CID 91884754
IUPAC Name diazanium;3-ethyl-2-[(E)-(3-ethyl-6-sulfonato-1,3-benzothiazol-2-ylidene)hydrazinylidene]-1,3-benzothiazole-6-sulfonate
SMILES [NH4+].[NH4+].CCN1\C(SC2=CC(=CC=C12)S([O-])(=O)=O)=N\N=C1/SC2=CC(=CC=C2N1CC)S([O-])(=O)=O

Specifications

Color Green
Quantity 1 g
Beilstein 7329461
Sensitivity Light sensitive
Solubility Information Dissolve in water at 50mg/ml. Solution may be slightly hazy.
Formula Weight 548.68
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM20L005 Certificate TypeCertificate of Analysis Date22/01/2025 Catalog NumberJ65535.03,J65535.06
Lot NumberY15K001 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberJ65535.03,J65535.06
Lot NumberX17K025 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog NumberJ65535.03,J65535.06
Lot NumberW27K046 Certificate TypeCertificate of Analysis Date07/10/2024 Catalog NumberJ65535.03,J65535.06
Lot NumberQ20K006 Certificate TypeCertificate of Analysis Date11/06/2024 Catalog NumberJ65535.03,J65535.06
compliance-icons
Product Identifier
  • 2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2,2'-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.