missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Azobis(2-methylpropionamidine) dihydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 10005650
Click to view available options
Quantity:
25 g
50 g
250 g
Packaging:
Plastic Bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2997-92-4
Molecular Formula C8H18N6·2HCl
Molecular Weight (g/mol) 271.19
MDL Number MFCD00142725
InChI Key LXEKPEMOWBOYRF-UHFFFAOYSA-N
Synonym aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap
PubChem CID 76344
IUPAC Name 2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride
SMILES CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Density 0.4200g/mL
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (on dry substance) (Argentometry)
Packaging Plastic Bottle
Quantity 50 g
UN Number 3226
Beilstein 04,IV,3379
Solubility Information Solubility in water: soluble.
Formula Weight 271.19
Percent Purity 98%
Physical Form Granules
Chemical Name or Material 2,2′-Azobis(2-methylpropionamidine) dihydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'