missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Azobis(2-methylpropionamidine) dihydrochloride, 98%

Product Code. 10218502 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
50 g
250 g
Packaging:
Plastic bottle
Unit Size:
250g
25g
50g
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

Product Code. 10218502

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

CAS: 2997-92-4 | C8H18N6·2HCl | 271.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2997-92-4
Molecular Formula C8H18N6·2HCl
Molecular Weight (g/mol) 271.19
MDL Number MFCD00142725
InChI Key LXEKPEMOWBOYRF-UHFFFAOYSA-N
Synonym aaph, 2,2'-diazene-1,2-diyl bis 2-methylpropanimidamide dihydrochloride, 2,2'-azobis 2-methylpropionamidine dihydrochloride, ms 1 catalyst, 2,2'-azobis 2-amidinopropane dihydrochloride, unii-7381jdr72f, ccris 7279, ms 1, propanimidamide, 2,2'-azobis 2-methyl-, dihydrochloride, abap
PubChem CID 76344
IUPAC Name 2-[(1-amino-1-imino-2-methylpropan-2-yl)diazenyl]-2-methylpropanimidamide;dihydrochloride
SMILES CC(C)(C(=N)N)N=NC(C)(C)C(=N)N.Cl.Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Density 0.4200g/mL
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (on dry substance) (Argentometry)
Packaging Plastic bottle
Quantity 25 g
Beilstein 04,IV,3379
Solubility Information Solubility in water: soluble.
Formula Weight 271.19
Percent Purity 98%
Physical Form Granules
Chemical Name or Material 2,2′-Azobis(2-methylpropionamidine) dihydrochloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2,2''-Azobis(2-methylpropionamidine) dihydrochloride
Signal Word
  • Danger
Hazard Category
  • Self-heating substance Category 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H251-Self-heating: may catch fire.
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P235+P410-Keep cool. Protect from sunlight.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.