missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(+/-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
(±)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl plays an essential role in the organic synthesis of enantioselective transformations catalyzed by the complexes of ruthenium, rhodium and palladium. It is also employed in palladium-catalyzed arylamine coupling in the preparation of demethylthiocholchines. It is used with Cu(II) to catalyze the addition of arylsulfonamides to styrenes and olefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
(^+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl plays an essential role in the organic synthesis of enantioselective transformations catalyzed by the complexes of ruthenium, rhodium and palladium. It is also employed in palladium-catalyzed arylamine coupling in the preparation of demethylthiocholchines. It is used with Cu(II) to catalyze the addition of arylsulfonamides to styrenes and olefins.
Solubility
Soluble in tetrahydrofuran, benzene and dichloromethane. Slightly soluble in ether, methanol and ethanol. Insoluble in water.
Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
(^+)-2,2′-Bis(diphenylphosphino)-1,1′-binaphthyl plays an essential role in the organic synthesis of enantioselective transformations catalyzed by the complexes of ruthenium, rhodium and palladium. It is also employed in palladium-catalyzed arylamine coupling in the preparation of demethylthiocholchines. It is used with Cu(II) to catalyze the addition of arylsulfonamides to styrenes and olefins.
Solubility
Soluble in tetrahydrofuran, benzene and dichloromethane. Slightly soluble in ether, methanol and ethanol. Insoluble in water.
Notes
Air sensitive. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Specifications
Melting Point | 281°C to 285°C |
Quantity | 100 g |
Beilstein | 5321443 |
Merck Index | 14,1223 |
Solubility Information | Soluble in tetrahydrofuran,benzene and dichloromethane. Slightly soluble in ether,methanol and ethanol. Insoluble in water. |
Formula Weight | 622.69 |
Percent Purity | 98% |
Chemical Name or Material | (+/-)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'