missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, 98%, Thermo Scientific Chemicals
116.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Use promo code "22914" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Absorbance | (0.1M in H2O) |
CAS Min % | 97.5 |
CAS Max % | 100.0 |
Melting Point | 103.0°C to 104.0°C |
Color | White to Brown |
Loss on Drying | 0.2% max. (80°C, 5 hrs) |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% |
Packaging | Plastic bottle |
Linear Formula | (HOCH2CH2)2NC(CH2OH)3 |
Show More |
Safety and Handling
Product Identifier
- 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol
- Warning
- Serious eye damage/eye irritation Category 2
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H290-May be corrosive to metals.
- P234-Keep only in original container.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P390-Absorb spillage to prevent material damage.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'